Skip to Content
Merck
All Photos(1)

Key Documents

87725

Sigma-Aldrich

Tetramethylammonium hexafluorophosphate

≥98.0% (gravimetric)

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$145.00

$145.00


Available to ship on14 April 2025Details


Request a Bulk Order

Select a Size

Change View
25 G
$145.00

About This Item

Linear Formula:
(CH3)4N(PF6)
CAS Number:
Molecular Weight:
219.11
Beilstein:
3730394
EC Number:
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

$145.00


Available to ship on14 April 2025Details


Request a Bulk Order

Quality Level

Assay

≥98.0% (gravimetric)

form

powder

mp

>300 °C (lit.)

SMILES string

C[N+](C)(C)C.F[P-](F)(F)(F)(F)F

InChI

1S/C4H12N.F6P/c1-5(2,3)4;1-7(2,3,4,5)6/h1-4H3;/q+1;-1

InChI key

ZFKULDQWLXAKIM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tobias Garling et al.
Journal of physics. Condensed matter : an Institute of Physics journal, 29(41), 414002-414002 (2017-07-21)
Surfaces whose macroscopic properties can be switched by light are potentially useful in a wide variety of applications. One such promising application is electrochemical sensors that can be gated by optically switching the electrode on or off. One way to
Nihed Rahmouni et al.
International journal of biological macromolecules, 113, 623-630 (2018-02-25)
Chitosan solubility and its antibacterial activity have been improved first of all with a chemical synthesis of N-benzimidazole-O-acetyl-chitosan in acetic medium and foremost through a potentiodynamic electrodeposition of this derivative. An association of one benzimidazole and three acetyl moieties per
Ludmila Šimková et al.
Journal of magnetic resonance (San Diego, Calif. : 1997), 323, 106895-106895 (2021-01-12)
The molecule of 2,2-dinitroethene-1,1-diamine (FOX-7) is one of the most interesting molecules with multiple redox centres stabilized by push-pull effect. To reveal the detailed mechanism of its electrochemical process radical intermediates formed in the course of its electrochemical reduction in

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service