Skip to Content
Merck
All Photos(1)

Key Documents

86718

Sigma-Aldrich

N,N,N′,N′-Tetraethylsulfamide

≥99.0%

Synonym(s):

TES

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C2H5)2NSO2N(C2H5)2
CAS Number:
Molecular Weight:
208.32
Beilstein:
1773566
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥99.0%

form

liquid

refractive index

n20/D 1.448 (lit.)
n20/D 1.449

bp

249-251 °C (lit.)

density

1.03 g/mL at 25 °C (lit.)

SMILES string

CCN(CC)S(=O)(=O)N(CC)CC

InChI

1S/C8H20N2O2S/c1-5-9(6-2)13(11,12)10(7-3)8-4/h5-8H2,1-4H3

InChI key

IYIAWAACGTUPCC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Aprotic, dipolar solvent for Grignard reactions

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

H.G. Richey et al.
The Journal of Organic Chemistry, 46, 2823-2823 (1981)
H.G. Richey et al.
The Journal of Organic Chemistry, 52, 479-479 (1987)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service