861804
8-(Diethylamino)octyl-3,4,5-trimethoxybenzoate hydrochloride
97%
Synonym(s):
TMB 8 hydrochloride, TMB-8 · HCl
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
(CH3O)3C6H2CO2(CH2)8N(C2H5)2 · HCl
CAS Number:
Molecular Weight:
431.99
Beilstein:
5466611
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Assay
97%
form
powder
mp
94-97 °C (lit.)
SMILES string
Cl[H].CCN(CC)CCCCCCCCOC(=O)c1cc(OC)c(OC)c(OC)c1
InChI
1S/C22H37NO5.ClH/c1-6-23(7-2)14-12-10-8-9-11-13-15-28-22(24)18-16-19(25-3)21(27-5)20(17-18)26-4;/h16-17H,6-15H2,1-5H3;1H
InChI key
KFJZVXKPPQIYCG-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
- Pharmacologically active inhibitor of hepatitis C virus life cycle
replaced by
Product No.
Description
Pricing
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Patrycja Libako et al.
Magnesium research, 25(1), 12-20 (2012-03-30)
Divalent cations, especially calcium and magnesium, have been shown to play an important regulatory role in endothelial and immune cells. To learn more about the interaction of these two metals in the regulation of cell growth, we altered the calcium/magnesium
Daniel Floryk et al.
Cancer research, 65(24), 11588-11596 (2005-12-17)
Recently, we have reported that inosine 5'-monophosphate dehydrogenase inhibitors, such as mycophenolic acid (MPA), induce the differentiation of PC-3 cells, which are derived from a human androgen-independent prostate cancer, into cells with a phenotype resembling maturing prostate secretory cells. Here
Hai-Long Wang et al.
The Chinese journal of physiology, 55(2), 101-107 (2012-05-09)
The purpose of this study was to determine the effects of an extract from Moringa oleifera (MO) on the development of monocrotaline (MCT)-induced pulmonary hypertension (PH) in Wistar rats. An ethanol extraction was performed on dried MO leaves, and HPLC
Nancy S Krieger et al.
American journal of physiology. Renal physiology, 300(1), F91-F97 (2010-11-05)
In vivo chronic metabolic acidosis induces net Ca2+ efflux from bone, and incubation of neonatal mouse calvariae in medium simulating physiological metabolic acidosis induces bone resorption. It appears that activation of the proton (H+) receptor OGR1 in the osteoblast leads
Kun-Ming Chen et al.
Protoplasma, 233(1-2), 39-49 (2008-08-30)
Two potent drugs, neomycin and TMB-8, which can block intracellular calcium release, were used to investigate their influence on pollen tube growth and cell wall deposition in Picea wilsonii. Apart from inhibiting pollen germination and pollen tube growth, the two
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service