Skip to Content
Merck
All Photos(1)

Key Documents

860700

Sigma-Aldrich

Z-Ser-OH

≥99%

Synonym(s):

Carbobenzyloxy-L-serine, Z-L-Serine

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$157.00

$157.00


Available to ship on22 April 2025Details


Request a Bulk Order

Select a Size

Change View
25 G
$157.00

About This Item

Linear Formula:
HOCH2CH(NHCO2CH2C6H5)CO2H
CAS Number:
Molecular Weight:
239.22
Beilstein:
2058314
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.22

$157.00


Available to ship on22 April 2025Details


Request a Bulk Order

Quality Level

Assay

≥99%

form

crystals

optical activity

[α]20/D +5.8°, c = 2.7 in acetic acid

reaction suitability

reaction type: solution phase peptide synthesis

mp

116-119 °C (lit.)

application(s)

peptide synthesis

SMILES string

OC[C@H](NC(=O)OCc1ccccc1)C(O)=O

InChI

1S/C11H13NO5/c13-6-9(10(14)15)12-11(16)17-7-8-4-2-1-3-5-8/h1-5,9,13H,6-7H2,(H,12,16)(H,14,15)/t9-/m0/s1

InChI key

GNIDSOFZAKMQAO-VIFPVBQESA-N

Looking for similar products? Visit Product Comparison Guide

Application

Building block in peptide synthesis; Starting material for the synthesis of various α-amino acids via the β-lactone

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jay M West et al.
PloS one, 7(8), e43877-e43877 (2012-09-07)
The mechanism of inactivation of human enzyme N-acylethanolamine-hydrolyzing acid amidase (hNAAA), with selected inhibitors identified in a novel fluorescent based assay developed for characterization of both reversible and irreversible inhibitors, was investigated kinetically and using matrix-assisted laser desorption/ionization time-of-flight mass
M Sypniewski et al.
The Journal of organic chemistry, 65(20), 6595-6600 (2000-10-29)
The enantiospecific synthesis of (R)-Boc-(Fmoc)-aminoglycine 7 was achieved. (S)-Cbz-serine 1 was reacted with diphenylphosphoryl azide in the presence of triethylamine to yield cyclic (S) carbamate 2. The ring nitrogen of 2 was protected with a Boc group (3). The cyclic
Jason Lejeune et al.
Biosensors & bioelectronics, 25(3), 604-608 (2009-03-27)
Changes detected in the imprinting effect by OMNiMIPs imprinted with multiple templates appear to be a function of the maximum template loading. Below the maximum template loading, the polymers imprinted with multiple compounds provide molecular recognition close to the polymers
Griet Van Zeebroeck et al.
Nature chemical biology, 5(1), 45-52 (2008-12-09)
Transporter-related nutrient sensors, called transceptors, mediate nutrient activation of signaling pathways through the plasma membrane. The mechanism of action of transporting and nontransporting transceptors is unknown. We have screened 319 amino acid analogs to identify compounds that act on Gap1

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service