Skip to Content
Merck
All Photos(2)

Key Documents

85971

Sigma-Aldrich

Dipyrrolidino(N-succinimidyloxy)carbenium hexafluorophosphate

≥98.0% (CHN)

Synonym(s):

N,N,N′,N′-Bis(tetramethylene)-O-(N-succinimidyl)uronium hexafluorophosphate, O-(N-Succinimidyl)-N,N,N′,N′-bis(tetramethylene)uronium hexafluorophosphate, HSPyU

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$112.00

$112.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$112.00

About This Item

Empirical Formula (Hill Notation):
C13H20F6N3O3P
CAS Number:
Molecular Weight:
411.28
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

$112.00


Check Cart for Availability

Request a Bulk Order

Assay

≥98.0% (CHN)

form

solid

reaction suitability

reaction type: Coupling Reactions

mp

143-153 °C

solubility

acetonitrile: 0.1 g/mL, clear

application(s)

peptide synthesis

storage temp.

2-8°C

SMILES string

F[P-](F)(F)(F)(F)F.O=C1CCC(=O)N1O\C(N2CCCC2)=[N+]3\CCCC3

InChI

1S/C13H20N3O3.F6P/c17-11-5-6-12(18)16(11)19-13(14-7-1-2-8-14)15-9-3-4-10-15;1-7(2,3,4,5)6/h1-10H2;/q+1;-1

InChI key

TVWKVGNCIJUXMY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

New, excellent and non-hygroscopic reagent for peptide coupling giving no cytotoxic reaction products
Reactant for synthesis of caspase sensitive gold nanoparticles

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Homan Kang et al.
Advanced materials (Deerfield Beach, Fla.), 32(6), e1905899-e1905899 (2019-12-20)
Advances in molecular imaging modalities have accelerated the diagnosis and treatment of human diseases. However, tumors less than 1 cm in size still remain difficult to localize by conventional means because of the difficulty in specific targeting/delivery to the tumor

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service