Skip to Content
Merck
All Photos(1)

Key Documents

856665

Sigma-Aldrich

N-Acetyl-Ala-Ala-Ala

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H19N3O5
CAS Number:
Molecular Weight:
273.29
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

Quality Level

SMILES string

C[C@H](NC(=O)[C@H](C)NC(=O)[C@H](C)NC(C)=O)C(O)=O

InChI

1S/C11H19N3O5/c1-5(12-8(4)15)9(16)13-6(2)10(17)14-7(3)11(18)19/h5-7H,1-4H3,(H,12,15)(H,13,16)(H,14,17)(H,18,19)/t5-,6-,7-/m0/s1

InChI key

DRYOODAJROGPQO-ACZMJKKPSA-N

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lanthanide probes in biological systems: the calcium binding site of pancreatic elastase as studied by terbium luminescence.
J de Jersey et al.
Biochemistry, 19(6), 1127-1132 (1980-03-18)
E R Vimr et al.
Journal of bacteriology, 153(3), 1252-1258 (1983-03-01)
Mutants of Salmonella typhimurium deficient in dipeptidyl carboxypeptidase have been isolated by screening for clones unable to use N-acetyl-L-alanyl-L-alanyl-L-alanine (AcAla3) as the sole nitrogen source. An insertion of the transposable element Tn10 near dcp (the locus coding for dipeptidyl carboxypeptidase)
Jan Kubelka et al.
Journal of the American Chemical Society, 124(19), 5325-5332 (2002-05-09)
Detailed spectral simulations based on ab initio density functional theory computations of the amide I and II infrared (IR) and vibrational circular dichroism (VCD) spectra for Ac-(Ala)(4)-NH(2), Ac-(Aib-Ala)(2)-NH(2), and Ac-(Aib)(4)-NH(2) constrained to 3(10)- and alpha-helical conformations are presented. Parameters from

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service