Skip to Content
Merck
All Photos(1)

Key Documents

855669

Sigma-Aldrich

L-Alanine ethyl ester hydrochloride

99%

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$41.30
25 G
$162.00

$41.30


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$41.30
25 G
$162.00

About This Item

Linear Formula:
CH3CH(NH2)COOC2H5 · HCl
CAS Number:
Molecular Weight:
153.61
Beilstein:
3594395
EC Number:
MDL number:
UNSPSC Code:
12352209
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.22

$41.30


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

99%

form

crystals

optical activity

[α]22/D +2.5°, c = 2.5 in H2O

reaction suitability

reaction type: solution phase peptide synthesis

mp

78-80 °C (dec.) (lit.)

application(s)

peptide synthesis

SMILES string

Cl[H].CCOC(=O)[C@H](C)N

InChI

1S/C5H11NO2.ClH/c1-3-8-5(7)4(2)6;/h4H,3,6H2,1-2H3;1H/t4-;/m0./s1

InChI key

JCXLZWMDXJFOOI-WCCKRBBISA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pavel L Padnya et al.
Organic & biomolecular chemistry, 13(21), 5894-5904 (2015-04-30)
New water-soluble p-tert-butylthiacalix[4]arenes containing peptide and quaternary ammonium fragments in cone and 1,3-alternate conformations were synthesized and characterized. The interaction of the macrocycles with DNA was studied by UV-spectroscopy, DLS and TEM. It was shown that the interaction of the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service