Skip to Content
Merck
All Photos(1)

Documents

852767

Sigma-Aldrich

5-Nitrouracil

98%

Synonym(s):

2,4-Dihydroxy-5-nitropyrimidine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H3N3O4
CAS Number:
Molecular Weight:
157.08
Beilstein:
10410
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

98%

form

powder

mp

>300 °C (lit.)

SMILES string

[O-][N+](=O)C1=CNC(=O)NC1=O

InChI

1S/C4H3N3O4/c8-3-2(7(10)11)1-5-4(9)6-3/h1H,(H2,5,6,8,9)

InChI key

TUARVSWVPPVUGS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Enzyme inhibitor.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

F H Lin et al.
Proceedings of the Society for Experimental Biology and Medicine. Society for Experimental Biology and Medicine (New York, N.Y.), 189(3), 353-361 (1988-12-01)
The kinetics of conversion of 5-fluoro-2'-deoxyuridine (FdUrd) to 5-fluorouracil (FUra) by isolated rat intestinal epithelial cells was investigated. Also, the effects of potential inhibitors of this reaction, which is catalyzed by uridine phosphorylase and thymidine phosphorylase, were determined. A 2.5%
S G Stepan'ian et al.
Biofizika, 34(5), 753-757 (1989-09-01)
The high-resolution IR-spectra of 5-nitrouracil and 5-bromouracil isolated in Ar matrices at 11 K were obtained for the first time. The conformational structure of uracil 5-substituents--thymine, 5-bromouracil, 5-nitrouracil--is calculated by the molecular mechanics and quantum--chemical MINDO/3 methods. The possibility of
X Gu et al.
Nucleic acids research, 24(6), 1059-1064 (1996-03-15)
tRNA in which uracil is completely replaced by 5-nitro-uracil was prepared by substituting 5-nitro-UTP for UTP in an in vitro transcription reaction. The rationale was that the 5-nitro substituent activates the 6-carbon of the Ura heterocycle towards nucleophiles, and hence
P J Nykänen et al.
APMIS : acta pathologica, microbiologica, et immunologica Scandinavica, 96(9), 768-772 (1988-09-01)
The degradation of 3H-thymidine under various cell culture conditions was analysed. It was found that a half of 3H-thymidine was degraded to 3H-thymine during 24 hours in PHA stimulation of blood lymphocytes. A control culture in which PHA was not
J A Smiley et al.
Analytical biochemistry, 266(1), 85-92 (1999-01-15)
A few organisms, notably some fungi, have the ability to metabolize thymidine to uracil, thus conserving the pyrimidine ring for subsequent metabolic use. Neurospora crassa possesses this pathway, termed the thymidine salvage pathway, and can utilize thymidine as a total

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service