Skip to Content
Merck
All Photos(1)

Key Documents

83980

Sigma-Aldrich

Rubidium chloride

purum p.a., ≥99.0% (AT)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
RbCl
CAS Number:
Molecular Weight:
120.92
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21
form:
crystals

grade

purum p.a.

Assay

≥99.0% (AT)

form

crystals

mp

715 °C (lit.)

density

2.8 g/mL at 25 °C (lit.)

anion traces

sulfate (SO42-): ≤200 mg/kg

cation traces

Ca: ≤50 mg/kg
Cd: ≤50 mg/kg
Co: ≤50 mg/kg
Cu: ≤50 mg/kg
Fe: ≤50 mg/kg
K: ≤5000 mg/kg
Na: ≤500 mg/kg
Ni: ≤50 mg/kg
Pb: ≤50 mg/kg
Zn: ≤50 mg/kg

SMILES string

[Cl-].[Rb+]

InChI

1S/ClH.Rb/h1H;/q;+1/p-1

InChI key

FGDZQCVHDSGLHJ-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Rubidium chloride can be used as:
  • An insulating layer in the OLED fabrication to improve the electroluminescent efficiency.
  • A component of an aqueous quaternary system in the study of its physicochemical properties using an isothermal evaporation method.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thermodynamics metastable phase equilibria of aqueous quaternary system LiCl+ KCl+ RbCl+ H2O at 323.15 K
Li Z, et al.
Fluid Phase Equilibria, 358, 131-136 (2013)
The effect of rubidium chloride on properties of organic light-emitting diodes
Lu Z, et al.
Solid-State Electron, 53(11), 1154-1158 (2009)
M L Główka et al.
Acta crystallographica. Section D, Biological crystallography, 61(Pt 4), 433-441 (2005-04-05)
Gramicidin D (gD) is a naturally occurring ionophoric antibiotic that forms membrane channels specific for monovalent cations. The crystal structure of the RbCl complex of gD has been determined at 1.14 A resolution from low-temperature (100 K) synchrotron-radiation data with
Ferenc Horkay et al.
The Journal of chemical physics, 125(23), 234904-234904 (2006-12-28)
The distribution of counterions in solutions of high molecular mass hyaluronic acid, in near-physiological conditions where mono- and divalent ions are simultaneously present, is studied by small angle neutron scattering and anomalous small angle x-ray scattering. The solutions contain either
Amer Alam et al.
Nature structural & molecular biology, 16(1), 30-34 (2008-12-23)
We report the crystal structure of the nonselective cation channel NaK from Bacillus cereus at a resolution of 1.6 A. The structure reveals the intracellular gate in an open state, as opposed to the closed form reported previously, making NaK

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service