Skip to Content
Merck
All Photos(1)

Key Documents

809810

Sigma-Aldrich

Clodinafop-propargyl-(phenoxy-13C6)

≥98 atom % 13C, ≥98% (CP)

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$1,180.00

$1,180.00


Estimated to ship on22 May 2025


Request more information

Select a Size

Change View
1 MG
$1,180.00

About This Item

Empirical Formula (Hill Notation):
13C6C11H13ClFNO4
Molecular Weight:
355.70
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.12

$1,180.00


Estimated to ship on22 May 2025


Request more information

isotopic purity

≥98 atom % 13C

Quality Level

Assay

≥98% (CP)

form

crystals

technique(s)

mass spectrometry (MS): suitable

mass shift

M+6

storage temp.

−20°C

SMILES string

C#CCOC(C(C)O[13C]([13CH]=[13CH]1)=[13CH][13CH]=[13C]1OC2=C(F)C=C(Cl)C=N2)=O

InChI

1S/C17H13ClFNO4/c1-3-8-22-17(21)11(2)23-13-4-6-14(7-5-13)24-16-15(19)9-12(18)10-20-16/h1,4-7,9-11H,8H2,2H3/i4+1,5+1,6+1,7+1,13+1,14+1

InChI key

JBDHZKLJNAIJNC-ANBMRESWSA-N

Related Categories

Signal Word

Warning

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Skin Sens. 1 - STOT RE 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service