Skip to Content
Merck
All Photos(1)

Documents

808717

Sigma-Aldrich

Titanium disulfide

Crystal, 99.995%

Synonym(s):

Titanium disulphide, Titanium sulfide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
TiS2
CAS Number:
Molecular Weight:
112.00
EC Number:
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:

Quality Level

Assay

99.995%

form

crystals

L × W × H

0.5 cm × 0.5 cm × 0.1-0.15 mm, typical size, some variation in dimensions may occur with different batches

SMILES string

S=[Ti]=S

InChI

1S/2S.Ti

InChI key

CFJRPNFOLVDFMJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

The two dimensional layered inorganic nanomaterials have drawn significant attention recently owing to their thickness dependent physical and chemical properties. They find applications in photocatalysis, phtotovoltaics, gas sensing, lithium ion batteries, field effect transistors, spintronics etc. Transition metal dischalcogenides represent an important class of inorganic 2D materials having the aforementioned applications.

Preparation Note

Store under nitrogen atmosphere.

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Self-heat. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

4.2 - Pyrophoric and self-heating hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Exploration of Nanostructured Functional Materials Based on Hybridization of Inorganic 2D Nanosheets.
Gunjakar JL, et al.
The Journal of Physical Chemistry C, 118, 3847-3863 (2014)
The super materials that could trump graphene.
Elizabeth Gibney
Nature, 522(7556), 274-276 (2015-06-19)
Sheneve Z Butler et al.
ACS nano, 7(4), 2898-2926 (2013-03-08)
Graphene's success has shown that it is possible to create stable, single and few-atom-thick layers of van der Waals materials, and also that these materials can exhibit fascinating and technologically useful properties. Here we review the state-of-the-art of 2D materials

Articles

Professors summarize recent 2D materials synthesis advancements and biosensing applications in various fields.

Advances in scalable synthesis and processing of two-dimensional materials

Research focuses on sustainable and cost-effective power generation systems to meet the growing demand for environmentally friendly energy sources.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service