Skip to Content
Merck
All Photos(1)

Key Documents

808598

Sigma-Aldrich

CLAmP

Synonym(s):

N-Methoxy-N-methyl-1H-pyrrole-1-carboxamide, Heller-Sarpong Reagent

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H10N2O2
Molecular Weight:
154.17
UNSPSC Code:
12352000
PubChem Substance ID:

form

liquid

refractive index

n/D 1.510

density

1.126 g/mL

storage temp.

2-8°C

SMILES string

O=C(N(OC)C)N1C=CC=C1

InChI

1S/C7H10N2O2/c1-8(11-2)7(10)9-5-3-4-6-9/h3-6H,1-2H3

InChI key

NLSJGNBOCVXWHS-UHFFFAOYSA-N

Application

The CLAmP (Carbonyl Linchpin N, O-dimethylhydroxylamine pyrrole) reagent, reported by Sarpong and Heller, is a useful building block for the efficient synthesis of unsymmetrical ketones. CLAmP shows high versatility based on compatibility with both highly reactive, thermally unstable nucleophiles as well as less reactive stable nucleophiles. It reacts with Grignard reagents to form the corresponding Weinreb amides.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

230.0 °F

Flash Point(C)

110 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

One-pot Unsymmetrical Ketone Synthesis Employing a Pyrrole-Bearing Formal Carbonyl Dication Linchpin Reagent.
Heller ST, et al.
Angewandte Chemie (International Edition in English), 54(34), 9839-9843 (2015)

Related Content

Professor Heller's research focuses on mild acylation reactions using carbonylimidazole-derived reagents.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service