Skip to Content
Merck
All Photos(2)

Key Documents

806366

Sigma-Aldrich

Poly(N-isopropyl acrylamide)

carboxylic acid, thiol

Synonym(s):

PNIPAAM, PNIPAM

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$777.00

$777.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$777.00

About This Item

Empirical Formula (Hill Notation):
C4H7O2(C6H11NO)nSH
UNSPSC Code:
12352100
NACRES:
NA.23

$777.00


Check Cart for Availability

Request a Bulk Order

Quality Level

form

powder

mol wt

Mn 6,000

PDI

≤1.2

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Application

Novel polymer for development of thermosensitive coated micro/nano materials[1] including thermoresponsive polymeric drug delivery systems.[2] The thiol functional group can be used to conjugate to a variety of biomolecules, as well as thiol-ene click chemistry.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chung JE, et al.
Colloids Surf., B9(37) (1997)
Wei HH, et al.
Biomaterials, 28(1), 9-9 (2007)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service