Skip to Content
Merck
All Photos(1)

Key Documents

806358

Sigma-Aldrich

Poly(D,L-lactide-co-glycolide) acid terminated

lactide:glycolide 75:25, Mw 6,000-10,000

Synonym(s):

PLGA

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$672.00

$672.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$672.00

About This Item

Empirical Formula (Hill Notation):
(C3H4O2)x(C2H2O2)y
CAS Number:
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.23

$672.00


Check Cart for Availability

Request a Bulk Order

Quality Level

form

pellets

mol wt

Mw 6,000-10,000

storage temp.

−20°C

SMILES string

O2C(C(=O)OC(C2=O)C)C.O1CC(=O)OCC1=O

InChI

1S/C6H8O4.C4H4O4/c1-3-5(7)10-4(2)6(8)9-3;5-3-1-7-4(6)2-8-3/h3-4H,1-2H3;1-2H2

InChI key

LCSKNASZPVZHEG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service