Skip to Content
Merck
All Photos(1)

Key Documents

806110

Sigma-Aldrich

bis-MPA-Azide dendrimer

trimethylol propane core, generation 5

Synonym(s):

G5-TMP-Azide

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$968.00

$968.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
$968.00

About This Item

Empirical Formula (Hill Notation):
C1047H1622N288O378
Molecular Weight:
24291.78
UNSPSC Code:
12162002
NACRES:
NA.23

$968.00


Check Cart for Availability

Request a Bulk Order

Quality Level

form

(Liquid, solid, or semi-solid)

storage temp.

−20°C

General description

  • End Group Functionality: Azide surface group
  • Generation: 5
  • No. Surface Groups: 96
  • Calculated Mol. Wt.: 24292.1 g/mol
  • Other Notes: bis-MPA = 2,2-bis(hydroxymethyl)propionic acid

Application

Nontoxic dendrimers for drug delivery, with functional groups for conjugation or additional modifications.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anna Carlmark et al.
Chemical Society reviews, 42(13), 5858-5879 (2013-05-01)
Dendritic polymers are highly branched, globular architectures with multiple representations of functional groups. These nanoscale organic frameworks continue to fascinate researchers worldwide and are today under intensive investigation in application-driven research. A large number of potential application areas have been

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service