Skip to Content
Merck
All Photos(2)

Key Documents

804916

Sigma-Aldrich

trans-2-(4-Bromophenyl)vinylboronic acid MIDA ester

Synonym(s):

MIDA084

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$94.30

$94.30


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$94.30

About This Item

Empirical Formula (Hill Notation):
C13H13BBrNO4
Molecular Weight:
337.96
UNSPSC Code:
12352103
NACRES:
NA.22

$94.30


Check Cart for Availability

Request a Bulk Order

form

powder

Quality Level

mp

218-223 °C

SMILES string

CN1CC(=O)OB(OC(=O)C1)\C=C\c2ccc(Br)cc2

InChI

1S/C13H13BBrNO4/c1-16-8-12(17)19-14(20-13(18)9-16)7-6-10-2-4-11(15)5-3-10/h2-7H,8-9H2,1H3/b7-6+

InChI key

DGIULDYIMUHJCF-VOTSOKGWSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service