Skip to Content
Merck
All Photos(1)

Documents

804789

Sigma-Aldrich

2,2,2-Trifluoroethanol-d3

99 atom % D, 99% (CP)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3CD2OD
CAS Number:
Molecular Weight:
103.06
EC Number:
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.21

isotopic purity

99 atom % D

Quality Level

Assay

99% (CP)

form

liquid

technique(s)

NMR: suitable

refractive index

n20/D 1.3 (lit.)

bp

77-80 °C (lit.)

mp

−44 °C (lit.)

density

1.415 g/mL at 25 °C (lit.)

mass shift

M+3

SMILES string

[2H]OC([2H])([2H])C(F)(F)F

InChI

1S/C2H3F3O/c3-2(4,5)1-6/h6H,1H2/i1D2,6D

InChI key

RHQDFWAXVIIEBN-IDPMSXFZSA-N

Looking for similar products? Visit Product Comparison Guide

Recommended products

Check out ChemisTwin®, our brand new online portal for identity confirmation and quantification of NMR spectra. Learn more or reach out to us for a free trial.

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Dam. 1 - Flam. Liq. 3 - Repr. 1B - STOT RE 2 Inhalation

Target Organs

Blood

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

84.2 °F - closed cup

Flash Point(C)

29 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service