Skip to Content
Merck
All Photos(1)

Key Documents

804010

Sigma-Aldrich

Co(salen,t-Bu, t-Bu)Cl

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C36H52ClCoN2O2
Molecular Weight:
639.20
UNSPSC Code:
12352300
PubChem Substance ID:

form

powder

reaction suitability

reagent type: catalyst
core: cobalt

mp

250 °C

storage temp.

2-8°C

SMILES string

Cl[Co]1OC(C(C(C)(C)C)=CC(C(C)(C)C)=C2)=C2/C=N/[C@](CCCC3)([H])[C@@]3([H])/N=C/C(C=C(C(C)(C)C)C=C4C(C)(C)C)=C4O1

InChI

1S/C36H54N2O2.ClH.Co/c1-33(2,3)25-17-23(31(39)27(19-25)35(7,8)9)21-37-29-15-13-14-16-30(29)38-22-24-18-26(34(4,5)6)20-28(32(24)40)36(10,11)12;;/h17-22,29-30,39-40H,13-16H2,1-12H3;1H;/q;;+3/p-3/b37-21+,38-22+;;/t29-,30-;;/m0../s1

InChI key

DIPYCKRNYQWVPM-RUIQGICGSA-K

Application

When utilized with organosilanes, catalytic amounts of Co(salen,t-Bu, t-Bu)Cl effects the isomerization of terminal alkenes.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Steven W M Crossley et al.
Journal of the American Chemical Society, 136(48), 16788-16791 (2014-11-15)
Catalytic amounts of Co(Sal(tBu,tBu))Cl and organosilane irreversibly isomerize terminal alkenes by one position. The same catalysts effect cycloisomerization of dienes and retrocycloisomerization of strained rings. Strong Lewis bases like amines and imidazoles, and labile functionalities like epoxides, are tolerated.

Related Content

Shenvi lab repurposed Jacobsen’s cobalt chloride complex for positional isomerization of terminal alkenes via reversible hydrogen atom transfer.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service