Skip to Content
Merck
All Photos(1)

Key Documents

803200

Sigma-Aldrich

DTSSP (3,3′-dithiobis(sulfosuccinimidyl propionate))

Synonym(s):

DTSSP, bis(sulfosuccinimidyl) 3,3′-dithiobis(propionate)

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
$318.00

$318.00


Available to ship on15 April 2025Details


Request a Bulk Order

Select a Size

Change View
50 MG
$318.00

About This Item

Empirical Formula (Hill Notation):
C14H14N2O14S4Na2
CAS Number:
Molecular Weight:
608.51
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.22

$318.00


Available to ship on15 April 2025Details


Request a Bulk Order

Assay

≥80%

Quality Level

form

powder

mol wt

608.51

reaction suitability

reagent type: cross-linking reagent

storage condition

desiccated

solubility

soluble (>6mg/ml in water)

shipped in

ambient

storage temp.

2-8°C

SMILES string

O=C(CC1S(=O)([O-])=O)N(OC(CCSSCCC(ON2C(CC(S(=O)([O-])=O)C2=O)=O)=O)=O)C1=O.[Na+].[Na+]

InChI

1S/C14H16N2O14S4.2Na/c17-9-5-7(33(23,24)25)13(21)15(9)29-11(19)1-3-31-32-4-2-12(20)30-16-10(18)6-8(14(16)22)34(26,27)28;;/h7-8H,1-6H2,(H,23,24,25)(H,26,27,28);;/q;2*+1/p-2

InChI key

XRWWBWZHHSXBNC-UHFFFAOYSA-L

General description

DTSSP is 3,3′-Dithiobis(sulfosuccinimidylpropionate), which contains an amine-reactive N-hydroxysulfosuccinimide (sulfo-NHS) ester at each end of an 8-carbon spacer arm. Sulfo-NHS esters react with primary amines at pH 7-9 to form stable amide bonds, along with release of the N-hydroxysulfosuccinimide leaving group. Proteins, including antibodies, generally have several primary amines in the side chain of lysine (K) residues and the N-terminus of each polypeptide that are available as targets for sulfo-NHS-ester crosslinking reagents. DSS, the non-sulfonated analog of DTSSP is also available for applications that require a membrane-permeable crosslinker.

Application

  • Enhancing Ocular Antisense Oligonucleotide Delivery: Utilizing Cowpea Chlorotic Mottle Virus-like particles, this study incorporates DTSSP for the enhancement of antisense oligonucleotide delivery targeting posterior segment ocular diseases, demonstrating a novel approach in biocompatible drug delivery systems (Pretto et al., 2021).
  • Regulation of Integrin-Tetraspanin Interactions: DTSSP is utilized to stabilize interactions between integrins and the tetraspanin CD9, providing critical insights into cell adhesion and signaling mechanisms, relevant to cancer and other pathologies (Torres-Gómez et al., 2021).
  • Gene Delivery Platform Development: A study on adeno-associated viral vectors employs DTSSP for cross-linking to tune cellular tropisms and enhance gene delivery, showing potential for targeted gene therapy applications (Yoo et al., 2020).
  • Protein Interactor Identification: DTSSP is instrumental in identifying interactors of the Arabidopsis thaliana Plant Natriuretic Peptide (AtPNP-A) using mass spectrometry, aiding in the understanding of plant signaling pathways (Turek et al., 2020).
  • Protein Delivery via Nanoparticles: Research on poly(l-lysine)-grafted-poly(ethylene glycol) nanoparticles highlights the use of DTSSP for redox-responsive crosslinking, improving the delivery mechanisms for therapeutic proteins (Seaberg et al., 2020).

Features and Benefits

  • Reactive groups: sulfo-NHS ester (both ends)
  • Reactive towards: amino groups (primary amines)
  • Sulfo-NHS ester reacts rapidly with any primary amine-containing molecule
  • Disulfide bond in the spacer arm is readily cleaved by 10-50 mM DTT or TCEP at pH 8.5
  • Spacer arm also easily cleaved with reducing SDS-PAGE sample loading
  • Cleavable crosslinker allows separation of crosslinked products
  • Water-soluble; compare with DSP
  • Membrane-impermeable, allowing for cell surface labeling

Caution

This product is sensitive to moisture. The vial is packaged in a resealable bag with a desiccant to reduce exposure to moisture. After cold storage, equilibrate the vial to room temperature before opening to reduce condensation inside the vial. Make fresh solutions. Storage of stock solutions is not recommended. After use, return the vial to the resealable bag. Close the bag and store the product at the recommended temperature.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service