Skip to Content
Merck
All Photos(1)

Key Documents

803022

Sigma-Aldrich

5-Methyltetrahydrofolic acid-(glutamic acid-13C5)

99 atom % 13C, 95% (CP)

Synonym(s):

5-MethyltetrahydroVitamin B9-(glutamic acid-13C5)

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$3,040.00
5 MG
$13,040.00

$3,040.00


Available to ship on14 April 2025Details


Request more information

Select a Size

Change View
1 MG
$3,040.00
5 MG
$13,040.00

About This Item

Empirical Formula (Hill Notation):
13C5C15H25N7O6
Molecular Weight:
464.42
UNSPSC Code:
12352205
PubChem Substance ID:
NACRES:
NA.12

$3,040.00


Available to ship on14 April 2025Details


Request more information

isotopic purity

99 atom % 13C

Quality Level

Assay

95% (CP)

form

powder

technique(s)

mass spectrometry (MS): suitable

color

white to off-white

application(s)

detection

mass shift

M+5

storage temp.

−20°C

SMILES string

O[13C]([13C][13C][13C]([13C](O)=O)NC(C1=CC=C(NC[C@@H]2N(C)C(C(N=C(N)N3)=O)=C3NC2)C=C1)=O)=O

InChI

1S/C20H20N7O6/c1-27-12(9-23-16-15(27)18(31)26-20(21)25-16)8-22-11-4-2-10(3-5-11)17(30)24-13(19(32)33)6-7-14(28)29/h2-5,12,22H,8-9H2,1H3,(H,24,30)(H,28,29)(H,32,33)(H4,21,23,25,26,31)/t12-/m0/s1/i6+1,7+1,13+1,14+1,19+1

InChI key

HMSZVEVPXBVRSZ-XCGRYNQXSA-N

Related Categories

General description

5-Methyltetrahydrofolic acid (5-MTHF) is an active folate metabolite. It plays an important role in the synthesis of methionine from homocysteine by donating methyl group.[1]
5-Methyltetrahydrofolic acid-(glutamic acid-13C5) is an isotope of vitamin B9 wherein all the five glutamate carbons are replaced by 13C6 carbon.

Application

5-Methyltetrahydrofolic acid-(glutamic acid-13C5) can be used as a stable isotope internal standard for accurate data interpretation in citrus juice samples by LC-MS.[2]

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of 5-Methyltetrahydrofolic Acid and Folic Acid in Citrus Juices Using Stable Isotope Dilution- Mass Spectrometry
Thomas PM, et al.
Journal of Agricultural and Food Chemistry, 51(5), 1293-1296 (2003)
Protein and Amino Acid Metabolism
Essentials of Medical Biochemistry: With Clinical Cases, 227-268 (2015)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service