Skip to Content
Merck
All Photos(1)

Key Documents

797421

Sigma-Aldrich

2-Boc-8-amino-5-oxa-2-azaspiro[3.4]octane

95%

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$2.40

$2.40


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
$2.40

About This Item

Empirical Formula (Hill Notation):
C11H20N2O3
CAS Number:
Molecular Weight:
228.29
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$2.40


Check Cart for Availability

Request a Bulk Order

Assay

95%

form

viscous liquid

functional group

ether

storage temp.

2-8°C

SMILES string

NC(CCO1)C21CN(C(OC(C)(C)C)=O)C2

InChI

1S/C11H20N2O3/c1-10(2,3)16-9(14)13-6-11(7-13)8(12)4-5-15-11/h8H,4-7,12H2,1-3H3

InChI key

LSACGXFQGVBFLY-UHFFFAOYSA-N

Application

Spirocyclic building block was first synthesized by Carreira and coworkers, which provides a new area of chemical space with straightforward functional handles for further diversification.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Spirocyclic modules containing four-membered rings are currently of growing interest to discovery chemists.

Related Content

Carreira Research & SpiroChem innovate spirocyclic building blocks, enhancing drug discovery with diverse functionalization sites and compact modules.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service