Skip to Content
Merck
All Photos(2)

Key Documents

796891

Sigma-Aldrich

3,5-Dimethyl-4H-1,2,4-triazole

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$380.00

$380.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$380.00

About This Item

Empirical Formula (Hill Notation):
C4H7N3
CAS Number:
Molecular Weight:
97.12
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$380.00


Check Cart for Availability

Request a Bulk Order

Assay

95%

Quality Level

form

solid

mp

142-146 °C (exp)

SMILES string

CC1=NN=C(C)N1

InChI

1S/C4H7N3/c1-3-5-4(2)7-6-3/h1-2H3,(H,5,6,7)

InChI key

XYYXDARQOHWBPO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jia Zhang et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 24(56), 15013-15018 (2018-07-11)
We characterized the T2 -exchange (T2ex ) magnetic resonance imaging (MRI) contrast of azole protons that have large chemical shifts from the water proton resonance as a function of pH, temperature, and chemical modification. Our results showed that 1,2,4-triazoles could

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service