Skip to Content
Merck
All Photos(1)

Key Documents

78275

Sigma-Aldrich

Methyl (R)-3-phenylbutyrate

puriss., ≥99.0% (sum of enantiomers, GC)

Synonym(s):

Methyl (R)-β-methylhydrocinnamate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H14O2
CAS Number:
Molecular Weight:
178.23
Beilstein:
3198978
MDL number:
UNSPSC Code:
12352000
PubChem Substance ID:

grade

puriss.

Assay

≥99.0% (sum of enantiomers, GC)

optical activity

[α]20/D −44±1°, c = 1% in benzene

refractive index

n20/D 1.497

density

1.020 g/mL at 20 °C (lit.)

SMILES string

COC(=O)C[C@@H](C)c1ccccc1

Other Notes

Chiral standard. Chiral building block used for the synthesis of the corresponding alcohol, amide and olefin

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R. Bussas et al.
The Journal of Organic Chemistry, 48, 2828-2828 (1983)
D.J. Cram
Journal of the American Chemical Society, 74, 2137-2137 (1952)
T. Kogure et al.
The Journal of Organic Chemistry, 49, 576-576 (1984)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service