Skip to Content
Merck
All Photos(1)

Key Documents

78060

Sigma-Aldrich

L-Phenylalanine benzyl ester hydrochloride

≥99.0% (AT)

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$124.00

$124.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$124.00

About This Item

Linear Formula:
C6H5CH2CH(NH2)COOCH2C6H5 · HCl
CAS Number:
Molecular Weight:
291.77
Beilstein:
3919200
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.22

$124.00


Check Cart for Availability

Request a Bulk Order

Assay

≥99.0% (AT)

form

crystals

optical activity

[α]20/D −13±1°, c = 1% in acetic acid: water (4:1)

reaction suitability

reaction type: solution phase peptide synthesis

mp

197-200 °C

application(s)

peptide synthesis

SMILES string

Cl.N[C@@H](Cc1ccccc1)C(=O)OCc2ccccc2

InChI

1S/C16H17NO2.ClH/c17-15(11-13-7-3-1-4-8-13)16(18)19-12-14-9-5-2-6-10-14;/h1-10,15H,11-12,17H2;1H/t15-;/m0./s1

InChI key

CEXFHIYDTRNBJD-RSAXXLAASA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service