779059
1-Azido-2-iodobenzene solution
~0.25 M in tert-butyl methyl ether, ≥97% (HPLC)
Synonym(s):
2-Iodophenyl azide solution
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C6H4IN3
CAS Number:
Molecular Weight:
245.02
Beilstein:
1941884
MDL number:
UNSPSC Code:
12352200
Assay
≥97% (HPLC)
concentration
~0.25 M in tert-butyl methyl ether
impurities
≤2.0% water
storage temp.
−20°C
SMILES string
Ic1ccccc1N=[N+]=[N-]
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT RE 1
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
-27.4 °F
Flash Point(C)
-33 °C
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service