Skip to Content
Merck
All Photos(2)

Key Documents

777625

Sigma-Aldrich

1-Amino-2-methyl-2-propanol

95% anhydrous basis

Synonym(s):

1-Amino-2-methylpropan-2-ol, 1,1-Dimethylethanolamine, 2-Amino-α,α-dimethylethanol, 2-Hydroxy-2-methyl-1-propylamine, 2-Hydroxyisobutylamine, 2-Methyl-2-hydroxypropylamine, 3-Amino-2-methyl-2-propanol, NSC 17697

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$100.00

$100.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$100.00

About This Item

Empirical Formula (Hill Notation):
C4H11NO
CAS Number:
Molecular Weight:
89.14
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$100.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

95% anhydrous basis

form

liquid

refractive index

n20/D 1.448

density

0.957 g/mL at 25 °C

functional group

amine
hydroxyl

storage temp.

2-8°C

SMILES string

CC(C)(O)CN

InChI

1S/C4H11NO/c1-4(2,6)3-5/h6H,3,5H2,1-2H3

InChI key

LXQMHOKEXZETKB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

May contain up to 10% water.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

164.8 °F - closed cup

Flash Point(C)

73.8 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sandi A Kwee et al.
Journal of nuclear medicine : official publication, Society of Nuclear Medicine, 55(6), 905-910 (2014-03-29)
This study investigated the prognostic significance of metabolically active tumor volume (MATV) measurements applied to (18)F-fluorocholine PET/CT in castration-resistant prostate cancer (CRPC). (18)F-fluorocholine PET/CT imaging was performed on 30 patients with CRPC. Metastatic disease was quantified on the basis of
Ernst Pittenauer et al.
Analytical and bioanalytical chemistry, 407(17), 5079-5089 (2015-01-31)
A new type of low-mass substituted 4-oxazolin product ions of [M + H](+) precursor ions of aminophospholipids (glycerophosphatidylethanolamine, glycerophosphatidyl-N-methylethanolamine, glycerophosphatidyl-N,N-dimethylethanolamine, glycerophosphatidylserine) resulting from high-energy collision-induced dissociation (matrix-assisted laser desorption/ionization time-of-flight/reflectron time-of-flight mass spectrometry) and low-energy collision-induced dissociation (e.g., electrospray ionization
Alexander R Parent et al.
Dalton transactions (Cambridge, England : 2003), 43(33), 12501-12513 (2014-07-09)
Iron tris(2-methylpyridyl)amine (TPA) and iron 1-(bis(2-methylpyridyl)amino)-2-methyl-2-propanoate (BPyA) salts are characterized as water oxidation catalysts (WOCs) using sodium periodate. Under the conditions used, these complexes serve as homogeneous WOCs as demonstrated via kinetic analysis and dynamic light scattering (DLS). The Fe(BPyA)
Anita Aranyi et al.
Journal of chromatography. A, 1387, 123-133 (2015-02-18)
The direct separation of the enantiomers of 1-(α-aminoarylmethyl)-2-naphthol, 1-(α-aminoalkyl)-2-naphthol, 2-(α-aminoarylmethyl)-1-naphthol analogues and 2-(1-amino-2-methylpropyl)-1-naphthol) was investigated in supercritical fluid chromatography. Five commercially available chiral stationary phases based on immobilized polysaccharide chiral selectors (Chiralpak IA, IB, IC, ID and IE) were evaluated.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service