Skip to Content
Merck
All Photos(1)

Key Documents

776793

Sigma-Aldrich

InP/ZnS quantum dots

stabilized with oleylamine ligands, fluorescence λem 560 nm, 5 mg/mL in toluene

Synonym(s):

Cadmium free core shell quantum dots, Cadmium free quantum dots, Core/Shell heavy metal free quantum dots, Fluorescent nanocrystals

Sign Into View Organizational & Contract Pricing

Select a Size

5 ML
$960.00

$960.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 ML
$960.00

About This Item

Linear Formula:
InP/ZnS
UNSPSC Code:
26111700
NACRES:
NA.23

$960.00


Check Cart for Availability

Request a Bulk Order

Quality Level

form

liquid

concentration

5 mg/mL in toluene

fluorescence

FWHM <85 nm, quantum yield ~25%
λem 560 nm

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

InP/ZnS core shell nanocrystals are Cadmium free/heavy metal free quantum dots suitable for different applications like light emitting diodes (LEDs; display; biomedical applications. The toxicity of Cd compounds
and the corresponding regulations worldwide makes these InP/ZnS quantum dots as more desirable alternatives for consumer applications.

Other Notes

Material has a shelf life of at least 1 year (stored at 4-10°C in the dark under inert gas such as nitrogen or argon)

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 3 - Asp. Tox. 1 - Carc. 1B - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT RE 2 - STOT SE 3

Target Organs

Central nervous system, Lungs, Respiratory system

WGK

WGK 3

Flash Point(F)

closed cup

Flash Point(C)

closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Patrick T K Chin et al.
Biomaterials, 31(26), 6823-6832 (2010-07-14)
Fluorescence molecular imaging is rapidly increasing its popularity in image guided surgery applications. To help develop its full surgical potential it remains a challenge to generate dual-emissive imaging agents that allow for combined visible assessment and sensitive camera based imaging.
Greco; T.
Proc. SPIE: Int. Soc. Opt. Eng., 8424(NanophotonicsIV, 842439/1-842439/1 (2012)
Arun Narayanaswamy et al.
ACS nano, 3(9), 2539-2546 (2009-08-18)
Visual color changes between 300 and 510 K were observed in the photoluminescence (PL) of colloidal InP/ZnS core-shell nanocrystals. A subsequent study of PL spectra in the range 2-510 K and fitting the temperature dependent line shift and line width

Articles

Dye-sensitized solar cells as a promising, low-cost photovoltaic technology.

Professor Sharma and colleagues review the synthesis and applications of this novel material. This includes a discussion of the unique properties of quantum dots and their suitability for solar cell applications, along with common synthesis techniques used to develop these materials.

Professor Xiaohu Gao (University of Washington, USA) provides a overview of recent quantum dot (QD) advancements and their potential for advancing bioassay and bioimaging technologies.

Perovskite quantum dots research progresses overcoming challenges, enabling rapid development of light-emitting devices.

See All

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service