Skip to Content
Merck
All Photos(2)

Key Documents

776599

Sigma-Aldrich

Fmoc-Gly-Tyr(tBu)-OH

97%

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
$316.00

$316.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
500 MG
$316.00

About This Item

Empirical Formula (Hill Notation):
C30H32N2O6
Molecular Weight:
516.58
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.26

$316.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

97%

form

powder or crystals

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

mp

168-171 °C

application(s)

peptide synthesis

functional group

Fmoc

storage temp.

2-8°C

SMILES string

OC([C@@H](NC(CNC(OCC1C2=C(C=CC=C2)C3=C1C=CC=C3)=O)=O)CC4=CC=C(OC(C)(C)C)C=C4)=O

InChI

1S/C30H32N2O6/c1-30(2,3)38-20-14-12-19(13-15-20)16-26(28(34)35)32-27(33)17-31-29(36)37-18-25-23-10-6-4-8-21(23)22-9-5-7-11-24(22)25/h4-15,25-26H,16-18H2,1-3H3,(H,31,36)(H,32,33)(H,34,35)/t26-/m0/s1

InChI key

SLKZAIKXJIWNHP-SANMLTNESA-N

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service