Skip to Content
Merck
All Photos(1)

Key Documents

776580

Sigma-Aldrich

Boc-Gly-Val-OH

95%

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$322.00

$322.00


Estimated to ship on01 May 2025


Request a Bulk Order

Select a Size

Change View
1 G
$322.00

About This Item

Empirical Formula (Hill Notation):
C12H22N2O5
CAS Number:
Molecular Weight:
274.31
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.22

$322.00


Estimated to ship on01 May 2025


Request a Bulk Order

Quality Level

Assay

95%

form

powder or crystals

reaction suitability

reaction type: Boc solid-phase peptide synthesis

color

white to beige

mp

109-113 °C

application(s)

peptide synthesis

storage temp.

2-8°C

SMILES string

OC([C@H](C(C)C)NC(CNC(OC(C)(C)C)=O)=O)=O

InChI

1S/C12H22N2O5/c1-7(2)9(10(16)17)14-8(15)6-13-11(18)19-12(3,4)5/h7,9H,6H2,1-5H3,(H,13,18)(H,14,15)(H,16,17)/t9-/m0/s1

InChI key

YXXMEXYAHMBYIU-VIFPVBQESA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service