Skip to Content
Merck
All Photos(1)

Key Documents

773638

Sigma-Aldrich

Poly(N,N-dimethylacrylamide), DDMAT terminated

average Mn 10,000, PDI ≤1.1

Synonym(s):

PDMAM, polyDMAM, polydimethylacrylamide methylpropionic acid dodecyltrithiocarbonate

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$330.00

$330.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$330.00

About This Item

Linear Formula:
HOCOCH(CH3)2(CH2CHCON(CH3)2)nSCS2C12H25
UNSPSC Code:
12352100
NACRES:
NA.23

$330.00


Check Cart for Availability

Request a Bulk Order

form

powder

Quality Level

mol wt

average Mn 10,000

mp

>200 °C

PDI

≤1.1

storage temp.

2-8°C

General description

Need help choosing the correct RAFT Agent? Please consult the RAFT Agent to Monomer compatibility table.

Application

Poly(N; N-dimethylacrylamide) is generally used in biological applications. Additionally; the alpha-chain end is a functional carboxylic acid (for potential specialty/coupling reactions). DDMAT is especially suited for the polymerization of styrene; acrylate; and acrylamide monomers to make lithographically and biologically important block copolymers. Useful as a chain Transfer Agent (CTA) or a functionalized polymer

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Reversible addition–fragmentation chain transfer (RAFT) polymerization is rapidly moving to the forefront in construction of drug and gene delivery vehicles.

Over the past two decades, the rapid advance of controlled living polymerization (CLP) techniques.

Microparticles in drug delivery: Study on controlling chitosan microparticle size and distribution, exploring encapsulation of BSA and TPP cross-linker.

The modification of biomacromolecules, such as peptides and proteins, through the attachment of synthetic polymers has led to a new family of highly advanced biomaterials with enhanced properties.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service