Skip to Content
Merck
All Photos(4)

Key Documents

772054

Sigma-Aldrich

Fmoc-D-Lys(Boc)-OH

98%, for peptide synthesis

Synonym(s):

Nα-Fmoc-N-ε-Boc-D-lysine

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$53.90
5 G
$90.90
25 G
$284.00

$53.90


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$53.90
5 G
$90.90
25 G
$284.00

About This Item

Empirical Formula (Hill Notation):
C26H32N2O6
CAS Number:
Molecular Weight:
468.54
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.26

$53.90


Check Cart for Availability

Request a Bulk Order

Product Name

Fmoc-D-Lys(Boc)-OH, 98%

Quality Level

Assay

98%

form

powder or crystals

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

mp

128-131 °C

application(s)

peptide synthesis

functional group

Boc
Fmoc

storage temp.

2-8°C

SMILES string

CC(C)(C)OC(=O)NCCCC[C@@H](NC(=O)OCC1c2ccccc2-c3ccccc13)C(O)=O

InChI

1S/C26H32N2O6/c1-26(2,3)34-24(31)27-15-9-8-14-22(23(29)30)28-25(32)33-16-21-19-12-6-4-10-17(19)18-11-5-7-13-20(18)21/h4-7,10-13,21-22H,8-9,14-16H2,1-3H3,(H,27,31)(H,28,32)(H,29,30)/t22-/m1/s1

InChI key

UMRUUWFGLGNQLI-JOCHJYFZSA-N

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nicholas J Kenyon et al.
PloS one, 8(10), e77730-e77730 (2013-11-10)
Nanocarriers can deliver a wide variety of drugs, target them to sites of interest, and protect them from degradation and inactivation by the body. They have the capacity to improve drug action and decrease undesirable systemic effects. We have previously
Si Chen et al.
Biomaterials, 117, 92-104 (2016-12-13)
In this work, mitochondria-targeting gold nanostar (AuNS) and anticarcinogen DOX were co-encapsulated in hyaluronic acid (HA) protective shell for tumor-targeting synergistic photothermal/chemo-therapy. Cationic peptide R
Jingfang Li et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 23(54), 13510-13517 (2017-07-12)
A series of cationic peptides with alternating hydrophilic and hydrophobic residues were elaborately designed and synthesized. These kinds of short peptides with protonated lysine groups can interact with anionic polyoxometalate nanoclusters through multivalent ionic bonds and hydrogen bonds, resulting in
Andreas Pech et al.
Nucleic acids research, 45(7), 3997-4005 (2017-02-06)
Biological evolution resulted in a homochiral world in which nucleic acids consist exclusively of d-nucleotides and proteins made by ribosomal translation of l-amino acids. From the perspective of synthetic biology, however, particularly anabolic enzymes that could build the mirror-image counterparts

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service