Skip to Content
Merck
All Photos(1)

Documents

771503

Sigma-Aldrich

Indeno[1,2-b]fluorene-6,12-dione

99%

Synonym(s):

trans-Fluorenacenedione, Indenofluorenedione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H10O2
CAS Number:
Molecular Weight:
282.29
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

99%

form

powder

mp

350-354 °C

SMILES string

O=C1c2ccccc2-c3cc4C(=O)c5ccccc5-c4cc13

InChI

1S/C20H10O2/c21-19-13-7-3-1-5-11(13)15-9-18-16(10-17(15)19)12-6-2-4-8-14(12)20(18)22/h1-10H

InChI key

AAUHGKXJBZEARK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Core unit affording n-type organic semiconductors for n-type FET
Indeno[1,2-b]fluorene-6,12-dione is a semiconducting polymer that can be used as an acceptor molecule in the fabrication of organic electronic devices such as organic light emitting diodes (OLEDs) and organic field effect transistors (OFETs).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Preparation, physical properties and n-type FET characteristics of substituted diindenopyrazinediones and bis (dicyanomethylene) derivatives
Nishida J, et al.
Journal of Materials Chemistry, 22(10), 4483-4490 (2012)
Preparation, physical properties and n-type FET characteristics of substituted diindenopyrazinediones and bis(dicyanomethylene) derivatives
Journal of Materials Chemistry, 22, 4483-4490 (2012)
Synthesis and optoelectronic properties of indeno (1, 2-b) fluorene-6, 12-dione donor-acceptor-donor triads
Frederickson CK and Haley MM
The Journal of Organic Chemistry, 79(22), 11241-11245 (2014)
An aromatic imine group enhances the EL efficiency and carrier transport properties of highly efficient blue emitter for OLEDs
Journal of Materials Chemistry, 20, 5930-5936 (2010)
High performance n-type field-effect transistors based on indenofluorenedione and diindenopyrazinedione derivatives
Nakagawa T, et al.
Chemistry of Materials, 20(8), 2615-2617 (2008)

Articles

Fabrication procedure of organic field effect transistor device using a soluble pentacene precursor.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service