Skip to Content
Merck
All Photos(1)

Key Documents

769886

Sigma-Aldrich

Resomer® LG 855 S, Poly(L-lactide-co-glycolide)

ester terminated, lactide:glycolide 85:15

Synonym(s):

Resomer® LG 855 S, PLGA

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$225.00
25 G
$748.00

$225.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$225.00
25 G
$748.00

About This Item

Linear Formula:
HO[C3H4O2]n[C2H2O2]mCH3
CAS Number:
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.23

$225.00


Check Cart for Availability

Request a Bulk Order

Quality Level

form

solid

feed ratio

lactide:glycolide 85:15

viscosity

2.5-3.5 dL/g

mp

159-166 °C

transition temp

Tg 53-63 °C

storage temp.

2-8°C

Application

Biodegradable, biocompatible, and bioresorbable copolymer composed of L-Lactide and Glycolide. This semi-crystalline material has been used in the fabrication of research medical devices and research tissue engineering solutions, such as orthopedic or soft tissue fixation devices. Degradation of this materials has been thoroughly studied and has been shown to be safely resorbed by the body after implantation. Modification of molecular weight and polymer composition allows for control of the degradation rate and mechanical stability of the polymer.

Legal Information

RESOMER is a registered trademark of Evonik Rohm GmbH

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. 분자량을 알고싶은데요 아무리 찾아봐도 알 수가 없네요

    1 answer
    1. The molecular weight for this product is approximately 450,000 g/mol.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service