Skip to Content
Merck
All Photos(1)

Documents

768642

Sigma-Aldrich

Poly(3,4-ethylenedioxythiophene)-poly(styrenesulfonate)

1.0 wt. % in H2O, high-conductivity grade

Synonym(s):

Orgacon HIL-1005, PEDOT:PSS, Poly(2,3-dihydrothieno-1,4-dioxin)-poly(styrenesulfonate)

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352103

grade

high-conductivity grade

form

liquid

concentration

1.0 wt. % in H2O

resistance

50-120 Ω/sq

refractive index

n20/D 1.341

pH

1.8-2.2

viscosity

7-12 mPa.s(22 °C) (typical)

density

1.002 g/mL at 25 °C

Looking for similar products? Visit Product Comparison Guide

General description

A conducting polymer such as poly(3,4-ethylenedioxythiophene) doped with poly(styrene sulfonate) anions (PEDOT/PSS) is widely used in various organic optoelectronic devices. PEDOT: PSS is a blend of cationic polythiopene derivative, doped with a polyanion. High electrical conductivity and good oxidation resistance of such polymers make it suitable for electromagnetic shielding and noise suppression. Thus, the polymer film was found to possess high transparency throughout the visible light spectrum and even into near IR and near UV regions, virtually 100% absorption from 900-2,000 nm. No absorption maximum from 400-800 nm. Impact of small electric and magnetic fields on the polymer was studied.
Poly(3,4-ethylenedioxythiophene)-poly(styrenesulfonate) (PEDOT:PSS) is a conductive polymer that is formed by electropolymerizing 3,4-ethylenedioxythiophene in a solution of poly(styrenesulfonate)(PSS). PEDOT is doped with positive ions and PSS with negative ions. It has the following properties that makes it a viable polymer in organic electronics:
  • low band gap
  • good optical properties
  • high conductivity
  • low redox potential
  • easy processing
  • tunable film forming ability

Application

Conductive ink based on the high conductivity grade PEDOT:PSS polymer dispersion. Suitable for deposition and patterning of transparent conductive films using slot die coating and spin coating in OPV application.
PEDOT:PSS can be used as a high conductivity material with a high charge mobility. It can be used for a wide range of energy based applications such as photodetectors, organic photovoltaics (OPV), dye sensitized solar cells (DSSCs), organic light emitting diodes (OLEDs), supercapacitors and other biomedical sensors.
Virtually 100% absorption from 900-2,000 nm. No absorption maximum from 400-800 nm. Conductive polymer blend.

Analysis Note

VLT (excl. substr.) : 80-85%
Work Function: 5.2 -5.4 eV
Coating thickness : 40μm wet
Drying temp. 130°C during 6 min.

Legal Information

Product of Agfa-Gevaert N.V.
Orgacon is a trademark of Agfa-Gevaert N.V.

replaced by

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Impact of unintentional oxygen doping on organic photodetectors
Euvrard J, et al.
Organic Electronics, 54(19), 64-71 (2018)
M. Hanyak, et al.
Journal of Applied Physics, 109, 074905-074905 (2011)
Keita Yasumoro et al.
Polymers, 12(2) (2020-02-23)
Polyethylene dioxythiophene and polyethylene sulfonic acid (PEDOT/PSS) composite is gathering attention as an organic transparent conductive film material. However, it requires a core-shell structure in which conductive PEDOT is covered with insulating PSS. Providing film formability and a carrier to
Roll-to-Roll Slot-Die Coated Organic Photovoltaic (OPV) Modules with High Geometrical Fill Factors
Galagan Y, et al.
Energy Technology, 3(8), 834-842 (2015)
High efficiency, fully inkjet printed organic solar cells with freedom of design
Eggenhuisen TM, et al.
Journal of Material Chemistry A, 3(14), 7255-7262 (2015)

Articles

A detailed article on conducting polymer materials for flexible organic photovoltaics (OPVs) applications.

Functional materials for printed electronics applications enable flexible displays, RFID tags, and biomedical sensors.

Progress in Organic Thermoelectric Materials & Devices including high ZT values of >0.2 at room temperature by p-type (PEDOT:PSS) & n-type (Poly[Kx(Ni-ett)]) materials are discussed.

The application of conducting polymers at the interface with biology is an exciting new trend in organic electronics research.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service