Skip to Content
Merck
All Photos(2)

Documents

768448

Sigma-Aldrich

Lithium aluminum hexahydride

hydrogen-storage grade, ≥95% (by XRD)

Synonym(s):

Lithium aluminum hydride, Lithium hexahydroaluminate, Trilithium aluminum hexahydride, trilithium hexahydroaluminate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Li3AlH6
CAS Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥95% (by XRD)

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

SMILES string

[Li+].[Li+].[Li+].[AlH6-3]

InChI

1S/Al.3Li.6H/q-3;3*+1;;;;;;

InChI key

OUPSVBUKCTVOLU-UHFFFAOYSA-N

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Find details here.

Application

Li-based complex hydrides; Li alanates are promising materials for hydrogen storage owing to their high H content and interesting desorption kinetic properties at moderate temps. Li3AlH6 is formed as an intermediate species during the decomposition of LiAlH4. Metal hydrides that can store high volumes of hydrogen at ambient conditions which can be released during combustion show promising applications in fuel cell propulsion systems.

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B - Water-react 2

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dilts; J. A.;
Inorganic Chemistry, 11(6), 1230-1230 (1972)
Chen, R.;
Materials Chemistry and Physics, 126(3), 989-989 (2011)
Jun Lu et al.
The journal of physical chemistry. B, 110(29), 14236-14239 (2006-07-21)
Complex metal hydrides are considered as a class of candidate materials for hydrogen storage. Lithium-based complex hydrides including lithium alanates (LiAlH(4) and Li(3)AlH(6)) are among the most promising materials owing to its high hydrogen content. In the present work, we

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service