Skip to Content
Merck
All Photos(2)

Key Documents

767727

Sigma-Aldrich

Chloro(5-methoxy-2-{1-[(4-methoxyphenyl)imino-N]ethyl}phenyl-C)(1,2,3,4,5-pentamethylcyclopentadienyl)iridium(III)

greener alternative

97%

Synonym(s):

Chloro[5-methoxy-2-[1-[(4-methoxyphenyl)imino-N]ethyl]phenyl- C][(1,2,3,4,5-η)-1,2,3,4,5-pentamethyl-2,4-cyclopenta dien-1-yl]iridium

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
$326.00

$326.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
500 MG
$326.00

About This Item

Empirical Formula (Hill Notation):
C26H31ClIrNO2
CAS Number:
Molecular Weight:
617.20
EC Number:
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

$326.00


Check Cart for Availability

Request a Bulk Order

Assay

97%

form

solid

reaction suitability

core: iridium
reagent type: catalyst

greener alternative product characteristics

Catalysis
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

impurities

~1% Dichloromethane

mp

200-239 °C

greener alternative category

SMILES string

[CH]1[CH][CH][CH][CH]1.COc2ccc(cc2)\N=C(/C)c3ccc(OC)cc3[Ir]Cl

InChI

1S/C16H16NO2.C5H5.ClH.Ir/c1-12(13-4-8-15(18-2)9-5-13)17-14-6-10-16(19-3)11-7-14;1-2-4-5-3-1;;/h4,6-11H,1-3H3;1-5H;1H;/q;;;+1/p-1/b17-12+;;;

InChI key

JZDNDPJBPBFFCT-DPSBJRLESA-M

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for catalytic efficiency. Click here for more information.

Application

Catalyst for reductive amination by transfer hydrogenation.

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Carc. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

A versatile catalyst for reductive amination by transfer hydrogenation.
Chao Wang et al.
Angewandte Chemie (International ed. in English), 49(41), 7548-7552 (2010-11-03)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service