Skip to Content
Merck
All Photos(1)

Key Documents

767271

Sigma-Aldrich

Polyester bis-MPA dendron, 16 hydroxyl, 1 allyl

generation 4

Synonym(s):

bisMPA dendrimer, bisMPA dendron

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12162002

description

hydroxyl surface groups

form

solid

mol wt

generation 4

feature

focal point allyl

no. Surface Groups

16

mp

308-313 °C

storage temp.

2-8°C

General description

calculated mol. wt. 1799.8

Application

Allyl dendrons can be coupled to thiol functional molecules

Other Notes

:Bis-MPA = 2,2-Bis(hydroxyl-methyl)propionic acid

Legal Information

Manufactured by Polymer Factory Sweden AB

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service