Skip to Content
Merck
All Photos(1)

Documents

767050

Sigma-Aldrich

2-(Acetylthio)acetic acid

≥90%

Synonym(s):

S-Acetyl-2-mercaptoacetic acid, S-Acetylthioglycolic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H6O3S
CAS Number:
Molecular Weight:
134.15
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥90%

form

liquid

refractive index

n20/D 1.506

density

1.298 g/mL at 25 °C

SMILES string

CC(=O)SCC(O)=O

InChI

1S/C4H6O3S/c1-3(5)8-2-4(6)7/h2H2,1H3,(H,6,7)

InChI key

QSBWDKUBOZHGOU-UHFFFAOYSA-N

Related Categories

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Puthupparampil V Scaria et al.
PloS one, 12(12), e0190312-e0190312 (2017-12-28)
Chemical conjugation of polysaccharide to carrier proteins has been a successful strategy to generate potent vaccines against bacterial pathogens. We developed a similar approach for poorly immunogenic malaria protein antigens. Our lead candidates in clinical trials are the malaria transmission
Oscar A Marcos-Contreras et al.
Journal of controlled release : official journal of the Controlled Release Society, 301, 54-61 (2019-03-16)
New advances in intra-arterial (IA) catheters offer clinically proven local interventions in the brain. Here we tested the effect of combining local IA delivery and vascular immunotargeting. Microinjection of tumor necrosis factor alpha (TNFα) in the brain parenchyma causes cerebral
Yi Yu et al.
Nature chemical biology, 16(4), 387-390 (2019-12-25)
Here, we report a rapid CRISPR-Cas9-mediated gene knock-in strategy that uses Cas9 ribonucleoprotein and 5'-modified double-stranded DNA donors with 50-base-pair homology arms and achieved unprecedented 65/40% knock-in rates for 0.7/2.5 kilobase inserts, respectively, in human embryonic kidney 293T cells. The identified

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service