Skip to Content
Merck
All Photos(1)

Key Documents

766526

Sigma-Aldrich

3-Allyloxy-1-Boc-azetidine

Synonym(s):

3-Allyloxyazetidine-1-carboxylic acid tert-butyl ester, tert-Butyl 3-allyloxyazetidine-1-carboxylate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H19NO3
CAS Number:
Molecular Weight:
213.27
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

form

liquid

refractive index

n20/D 1.455

bp

220-245 °C

density

0.996 g/mL at 25 °C

functional group

ether

storage temp.

2-8°C

SMILES string

CC(C)(C)OC(=O)N1CC(C1)OCC=C

InChI

1S/C11H19NO3/c1-5-6-14-9-7-12(8-9)10(13)15-11(2,3)4/h5,9H,1,6-8H2,2-4H3

InChI key

PZYUTLNRBHWRIR-UHFFFAOYSA-N

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service