Skip to Content
Merck
All Photos(1)

Key Documents

766445

Sigma-Aldrich

(Acetonitrile)[1,3-bis(2,6-diisopropylphenyl)imidazol-2-ylidene]gold(I) tetrafluoroborate

Synonym(s):

IPr Au(MeCN)BF4

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$132.00
500 MG
$465.00

$132.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
$132.00
500 MG
$465.00

About This Item

Empirical Formula (Hill Notation):
C29H39AuBF4N3
CAS Number:
Molecular Weight:
713.41
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

$132.00


Check Cart for Availability

Request a Bulk Order

form

solid

Quality Level

reaction suitability

core: gold
reagent type: catalyst

impurities

(May contain up to 15% of the chloride salt 696277.)

mp

162-167 °C (decomposition)

SMILES string

CC#N.F[B-](F)(F)F.CC(C)c1cccc(C(C)C)c1N2C=CN(C2=[Au+])c3c(cccc3C(C)C)C(C)C

InChI

1S/C27H36N2.C2H3N.Au.BF4/c1-18(2)22-11-9-12-23(19(3)4)26(22)28-15-16-29(17-28)27-24(20(5)6)13-10-14-25(27)21(7)8;1-2-3;;2-1(3,4)5/h9-16,18-21H,1-8H3;1H3;;/q;;+1;-1

InChI key

YZYFYLAQORXITI-UHFFFAOYSA-N

Application

Useful in the synthesis of pyrroles by amino-Claisen rearrangement

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service