Skip to Content
Merck
All Photos(1)

Key Documents

766410

Sigma-Aldrich

1-(2-Chlorophenyl)vinylboronic acid pinacol ester

Synonym(s):

2-[1-(2-Chlorophenyl)ethenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H18BClO2
CAS Number:
Molecular Weight:
264.56
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

form

solid

mp

33-39 °C

storage temp.

2-8°C

SMILES string

CC1(C)OB(OC1(C)C)C(=C)c2ccccc2Cl

InChI

1S/C14H18BClO2/c1-10(11-8-6-7-9-12(11)16)15-17-13(2,3)14(4,5)18-15/h6-9H,1H2,2-5H3

InChI key

LLYOIGXBMLPFMQ-UHFFFAOYSA-N

Application

Organoboron building blocks for cross coupling

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fang Gao et al.
Journal of the American Chemical Society, 132(32), 10961-10963 (2010-08-12)
A method for Ni-catalyzed hydroalumination of terminal alkynes, leading to the formation of alpha-vinylaluminum isomers efficiently (>98% conv in 2-12 h) and with high selectivity (95% to >98% alpha), is described. Catalytic alpha-selective hydroalumination reactions proceed in the presence of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service