Skip to Content
Merck
All Photos(1)

Key Documents

765244

Sigma-Aldrich

Lithium-7Li hexafluorophosphate

95 atom % 7Li, 98% (CP)

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$3,380.00

$3,380.00


Check Cart for Availability

Request more information

Select a Size

Change View
1 G
$3,380.00

About This Item

Linear Formula:
7LiPF6
Molecular Weight:
151.98
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.12

$3,380.00


Check Cart for Availability

Request more information

isotopic purity

95 atom % 7Li

Quality Level

Assay

98% (CP)

form

crystals

mass shift

depleted

SMILES string

F[P-](F)(F)(F)(F)F.[7Li+]

InChI

1S/F6P.Li/c1-7(2,3,4,5)6;/q-1;+1/i;1+0

InChI key

AXPLOJNSKRXQPA-QMVMUTFZSA-N

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Dam. 1 - Skin Corr. 1A - STOT RE 1 Inhalation

Target Organs

Bone,Teeth

Storage Class Code

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kewei Liu et al.
ACS nano, 9(6), 6041-6049 (2015-06-06)
The two-dimensional single-layer and few-layered graphene exhibit many attractive properties such as large specific surface area and high charge carrier mobility. However, graphene sheets tend to stack together and form aggregates, which do not possess the desirable properties associated with
M D S Lekgoathi et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 153, 651-654 (2015-10-11)
The structure of LiPF6 has been probed using Raman scattering as well as pXRD and the results are compared and contrasted. The conventional Bragg angle scattering pXRD determines that dry LiPF6 crystallizes in a trigonal structure (Space Group R-3 (148))
Shijia Zhao et al.
Nanoscale, 7(5), 1984-1993 (2014-12-30)
Hydrogenated carbon nanomaterials exhibit many advantages in both mechanical and electrochemical properties, and thus have a wide range of potential applications. However, methods to control the hydrogenation and the effect of hydrogenation on the microstructure and properties of the produced
Jiangfeng Qian et al.
Nature communications, 6, 6362-6362 (2015-02-24)
Lithium metal is an ideal battery anode. However, dendrite growth and limited Coulombic efficiency during cycling have prevented its practical application in rechargeable batteries. Herein, we report that the use of highly concentrated electrolytes composed of ether solvents and the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service