Skip to Content
Merck
All Photos(1)

Key Documents

764159

Sigma-Aldrich

1-(Bromomethyl)adamantane

97%

Synonym(s):

1-Adamantylmethyl bromide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H17Br
CAS Number:
Molecular Weight:
229.16
UNSPSC Code:
12352100

Assay

97%

form

solid

mp

40-45 °C

storage temp.

2-8°C

InChI

1S/C11H17Br/c12-7-11-4-8-1-9(5-11)3-10(2-8)6-11/h8-10H,1-7H2

InChI key

RLRBYCAVZBQANU-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service