Skip to Content
Merck
All Photos(2)

Key Documents

761494

Sigma-Aldrich

4-Bromo-1-naphthol

95%

Synonym(s):

1-Bromo-4-hydroxynaphthalene, 4-Bromo-1-naphthalenol

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$116.00

$116.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$116.00

About This Item

Empirical Formula (Hill Notation):
C10H7BrO
CAS Number:
Molecular Weight:
223.07
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$116.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

95%

form

solid

mp

128-133 °C

functional group

bromo

SMILES string

Oc1ccc(Br)c2ccccc12

InChI

1S/C10H7BrO/c11-9-5-6-10(12)8-4-2-1-3-7(8)9/h1-6,12H

InChI key

OUNQUWORSXHSJN-UHFFFAOYSA-N

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

257.0 °F - closed cup

Flash Point(C)

125 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marko Weimar et al.
The Journal of organic chemistry, 75(8), 2718-2721 (2010-03-23)
Starting from Dane's diene and methylcyclopentenedione, (+)-estrone is synthesized along the Quinkert-Dane route in 24% total yield. The key step is an enantioselective Diels-Alder reaction promoted by an amidinium catalyst as efficiently as by a traditional Ti-TADDOLate Lewis acid.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service