Skip to Content
Merck
All Photos(2)

Key Documents

758426

Sigma-Aldrich

4-Amino-3,5-dibromopyridine

95%

Synonym(s):

3,5-Dibromo-4-pyridinamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H4Br2N2
CAS Number:
Molecular Weight:
251.91
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
form:
solid
Assay:
95%

Assay

95%

form

solid

mp

165-170 °C

functional group

bromo

SMILES string

Nc1c(Br)cncc1Br

InChI

1S/C5H4Br2N2/c6-3-1-9-2-4(7)5(3)8/h1-2H,(H2,8,9)

InChI key

MNRBVTDLMUNVLD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service