Skip to Content
Merck
All Photos(1)

Key Documents

755923

Sigma-Aldrich

N-Methyl-3-aminooxetane

97%

Synonym(s):

N-Methyloxetan-3-amine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H9NO
CAS Number:
Molecular Weight:
87.12
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Quality Level

Assay

97%

form

liquid

refractive index

n20/D 1.443

bp

121.4 °C

density

0.979 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

CNC1COC1

InChI

1S/C4H9NO/c1-5-4-2-6-3-4/h4-5H,2-3H2,1H3

InChI key

JUIXJPRSYHSLHK-UHFFFAOYSA-N

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

104.0 °F

Flash Point(C)

40 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service