Skip to Content
Merck
All Photos(1)

Key Documents

755338

Sigma-Aldrich

4-Acetoxybenzyl alcohol

97%

Synonym(s):

4-(Acetyloxy)benzenemethanol, 4-(Hydroxymethyl)phenyl acetate

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$180.00

$180.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$180.00

About This Item

Empirical Formula (Hill Notation):
C9H10O3
CAS Number:
Molecular Weight:
166.17
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$180.00


Check Cart for Availability

Request a Bulk Order

Assay

97%

form

solid

refractive index

n20/D 1.529

mp

37-42 °C

density

1.169 g/mL at 25 °C

functional group

ester
hydroxyl

storage temp.

2-8°C

SMILES string

CC(=O)Oc1ccc(CO)cc1

InChI

1S/C9H10O3/c1-7(11)12-9-4-2-8(6-10)3-5-9/h2-5,10H,6H2,1H3

InChI key

FKMVNGWJGSSDCF-UHFFFAOYSA-N

Related Categories

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

230.0 °F

Flash Point(C)

110 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Joerg Isensee et al.
Journal of cell science, 130(13), 2134-2146 (2017-05-19)
Maturation of nociceptive neurons depends on changes in transcription factors, ion channels and neuropeptides. Mature nociceptors initiate pain in part by drastically reducing the activation threshold via intracellular sensitization signaling. Whether sensitization signaling also changes during development and aging remains
Frank Schwede et al.
Molecular endocrinology (Baltimore, Md.), 29(7), 988-1005 (2015-06-11)
cAMP-elevating agents such as the incretin hormone glucagon-like peptide-1 potentiate glucose-stimulated insulin secretion (GSIS) from pancreatic β-cells. However, a debate has existed since the 1970s concerning whether or not cAMP signaling is essential for glucose alone to stimulate insulin secretion.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service