Skip to Content
Merck
All Photos(1)

Documents

754013

Sigma-Aldrich

PFO-DBT

average Mw 10,000-50,000

Synonym(s):

Poly[2,7-(9,9-dioctylfluorene)-alt-4,7-bis(thiophen-2-yl)benzo-2,1,3-thiadiazole]

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C43H46N2S3)n
CAS Number:
UNSPSC Code:
12352103
NACRES:
NA.23

description

Band gap: 1.87 eV

form

solid

mol wt

average Mw 10,000-50,000

loss

0.5 wt. % TGA, 400 °C

transition temp

Tm >400 °C

Orbital energy

HOMO -5.4 eV 
LUMO -3.53  (CV)

OPV Device Performance

ITO/PEDOT:PSS/PFO-DPT:PC61BM (1:2)/Ba/Al

  • Short-circuit current density (Jsc): 5.18 mA/cm2
  • Open-circuit voltage (Voc): 0.95 V
  • Fill Factor (FF): 0.35
  • Power Conversion Efficiency (PCE): 2.24 %

semiconductor properties

P-type (mobility=3×10−4 cm2/V·s)

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fluorene-based low band-gap copolymers for high performance photovoltaic devices
Zhou, Q.; et al.
Applied Physics Letters, 84, 1653-1653 (2004)

Articles

The development of high-performance conjugated organic molecules and polymers has received widespread attention in industrial and academic research.

Organic materials in optoelectronic devices like LEDs and solar cells are of significant academic and commercial interest.

Organic photovoltaics (OPVs) represent a low-cost, lightweight, and scalable alternative to conventional solar cells. While significant progress has been made in the development of conventional bulk heterojunction cells, new approaches are required to achieve the performance and stability necessary to enable commercially successful OPVs.

Thin, lightweight, and flexible electronic devices meet widespread demand for scalable, portable, and robust technology.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service