Skip to Content
Merck
All Photos(2)

Key Documents

753602

Sigma-Aldrich

Tetrahydropyran-4-boronic acid pinacol ester

Synonym(s):

4,4,5,5-Tetramethyl-2-(tetrahydro-2H-pyran-4-yl)-1,3,2-dioxaborolane, Tetrahydro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-2H-pyran

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$352.00

$352.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$352.00

About This Item

Empirical Formula (Hill Notation):
C11H21BO3
CAS Number:
Molecular Weight:
212.09
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

$352.00


Check Cart for Availability

Request a Bulk Order

form

liquid

functional group

ether

storage temp.

2-8°C

SMILES string

CC(C(C)(C)O1)(C)OB1C2CCOCC2

InChI

1S/C11H21BO3/c1-10(2)11(3,4)15-12(14-10)9-5-7-13-8-6-9/h9H,5-8H2,1-4H3

InChI key

NLSMOSUUBUCSPL-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service