Skip to Content
Merck
All Photos(1)

Documents

751979

Sigma-Aldrich

Calcium iodide hydrate

99.995% trace metals basis

Synonym(s):

Calcium diiodide hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CaI2·xH2O
CAS Number:
Molecular Weight:
293.89 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

Assay

99.995% trace metals basis

form

solid

impurities

≤60.0 ppm Trace Metal Analysis

mp

783 °C (lit.)

SMILES string

O.I[Ca]I

InChI

1S/Ca.2HI.H2O/h;2*1H;1H2/q+2;;;/p-2

InChI key

XIRQETYMJRSWOH-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Calcium Iodide finds use in the preparation of complex used in polymerization and hydroamination† as well as serving as a matrix for luminescent materials.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jain, S. K.; et al.
Indian J. Chem. A, 38, 778-778 (1999)
Malcolm H Chisholm et al.
Inorganic chemistry, 43(21), 6717-6725 (2004-10-13)
A series of monomeric amide or aryloxide complexes of the form LCaX, where L = CH[CMeNC(6)H(3)-2,6-](2), CH[CMeNC(6)H(4)-2-OMe](2), a bulky tris-pyrazolyl borate, Tp(iPr) or Tp(tBu) or 9-BBN-pz(2) and X = N(SiMe(3))(2) or OC(6)H(3)-2,6-, has been prepared and characterized and investigated in
Novosad, S.S.; et al.
Inorganic Materials, 44, 900-904 (2008)
Mark R Crimmin et al.
Journal of the American Chemical Society, 131(28), 9670-9685 (2009-06-26)
The beta-diketiminate-stabilized calcium amide complex [{ArNC(Me)CHC(Me)NAr}Ca{N(SiMe(3))(2)}(THF)] (Ar = 2,6-diisopropylphenyl) and magnesium methyl complex [{ArNC(Me)CHC(Me)NAr}Mg(Me)(THF)] are reported as efficient precatalysts for hydroamination/cyclization of aminoalkenes. The reactions proceeded under mild conditions, allowing the synthesis of five-, six-, and seven-membered heterocyclic compounds. Qualitative

Articles

Three approaches generate white light, including LED-based down-conversion for broader applications.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service